Assay Details
| Gene Symbol: RECQ4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104773.7 | NP_176289.7 | 19 - 20 | 2855 | 147 | |
| GenBank mRNA | AJ404474.1 | - | 19 - 20 | 2856 | 147 | |
More Information
Set Membership: |
|

RECQ4B
Genomic Map