Assay Details
| Gene Symbol: TPS04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104793.3 | NP_564772.1 | 1 - 2 | 440 | 138 | |
| GenBank mRNA | AY059757.1 | - | 1 - 2 | 438 | 138 | |
| BT001960.1 | - | 1 - 2 | 386 | 138 | ||
More Information
Set Membership: |
|

TPS04
Genomic Map