Assay Details
| Gene Symbol: TPS04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104793.3 | NP_564772.1 | 2 - 3 | 642 | 91 | |
| GenBank mRNA | AY059757.1 | - | 2 - 3 | 640 | 91 | |
| BT001960.1 | - | 2 - 3 | 588 | 91 | ||
More Information
Set Membership: |
|

TPS04
Genomic Map