Assay Details
| Gene Symbol: TPS04 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104793.3 | NP_564772.1 | 3 - 4 | 874 | 146 | |
| GenBank mRNA | AY059757.1 | - | 3 - 4 | 872 | 146 | |
| BT001960.1 | - | 3 - 4 | 820 | 146 | ||
More Information
Set Membership: |
|

TPS04
Genomic Map