Assay Details
| Gene Symbol: EDA16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124052.2 | NP_001117524.1 | 6 - 7 | 1779 | 98 | |
| NM_001124053.2 | NP_001117525.1 | 6 - 7 | 2108 | 98 | ||
| NM_104795.4 | NP_176309.2 | 6 - 7 | 2080 | 98 | ||
More Information
Set Membership: |
|

EDA16
Genomic Map