Assay Details
| Gene Symbol: SYP124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333948.1 | NP_001319282.1 | 2 - 3 | 512 | 66 | |
| NM_104810.3 | NP_176324.1 | 1 - 2 | 660 | 66 | ||
| GenBank mRNA | DQ446382.1 | - | 1 - 2 | 477 | 66 | |
More Information
Set Membership: |
|

SYP124
Genomic Map