Assay Details
| Gene Symbol: TIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104839.3 | NP_564780.1 | 1 - 2 | 177 | 68 | |
| GenBank mRNA | AY136299.1 | - | 1 - 2 | 158 | 68 | |
| BT003431.1 | - | 1 - 2 | 115 | 68 | ||
More Information
Set Membership: |
|

TIM13
Genomic Map