Assay Details
| Gene Symbol: PTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104901.2 | NP_176411.2 | 4 - 5 | 996 | 66 | |
| GenBank mRNA | AY058836.1 | - | 4 - 5 | 996 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTR6| Gene Symbol: PTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104901.2 | NP_176411.2 | 4 - 5 | 996 | 66 | |
| GenBank mRNA | AY058836.1 | - | 4 - 5 | 996 | 66 | |
Set Membership: |
|