Assay Details
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104918.5 | NP_176428.1 | 2 - 3 | 645 | 97 | |
| GenBank mRNA | AK230351.1 | - | 2036 | 97 | ||
| AY045876.1 | - | 2 - 3 | 370 | 97 | ||
| AY062685.1 | - | 2 - 3 | 417 | 97 | ||
| AY093381.1 | - | 2 - 3 | 341 | 97 | ||
| AY133851.1 | - | 2 - 3 | 341 | 97 | ||
More Information
Set Membership: |
|

ACO2
Genomic Map