Assay Details
| Gene Symbol: FMO GS-OX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104935.2 | NP_564797.1 | 6 - 7 | 1600 | 88 | |
| GenBank mRNA | AY054642.1 | - | 6 - 7 | 1231 | 88 | |
| AY081554.1 | - | 6 - 7 | 1160 | 88 | ||
More Information
Set Membership: |
|

FMO GS-OX4
Genomic Map