Assay Details
| Gene Symbol: NOGC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334039.1 | NP_001321123.1 | 2 - 3 | 625 | 83 | |
| NM_001334040.1 | NP_001321124.1 | 2 - 3 | 661 | 83 | ||
| NM_104936.2 | NP_176446.1 | 2 - 3 | 708 | 83 | ||
More Information
Set Membership: |
|

NOGC1
Genomic Map