Assay Details
| Gene Symbol: NOGC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334039.1 | NP_001321123.1 | 6 - 7 | 1173 | 93 | |
| NM_104936.2 | NP_176446.1 | 6 - 7 | 1319 | 93 | ||
More Information
Set Membership: |
|

NOGC1
Genomic Map