Assay Details
| Gene Symbol: Hop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104951.4 | NP_176461.1 | 3 - 4 | 1354 | 96 | |
| GenBank mRNA | BT020538.1 | - | 3 - 4 | 1320 | 96 | |
More Information
Set Membership: |
|

Hop2
Genomic Map