Assay Details
| Gene Symbol: ASP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334052.1 | NP_001321076.1 | 1 - 2 | 100 | 79 | |
| NM_104958.2 | NP_564803.1 | 1 - 2 | 100 | 79 | ||
| NM_179507.2 | NP_849838.1 | 1 - 2 | 183 | 79 | ||
More Information
Set Membership: |
|

ASP4
Genomic Map