Assay Details
| Gene Symbol: ASP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334052.1 | NP_001321076.1 | 5 - 6 | 419 | 101 | |
| NM_104958.2 | NP_564803.1 | 5 - 6 | 413 | 101 | ||
| NM_179507.2 | NP_849838.1 | 5 - 6 | 502 | 101 | ||
More Information
Set Membership: |
|

ASP4
Genomic Map