Assay Details
| Gene Symbol: NRPD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198369.1 | NP_001185298.1 | 8 - 9 | 854 | 68 | |
| NM_001334061.1 | NP_001320709.1 | 7 - 8 | 1044 | 68 | ||
| NM_001334062.1 | NP_001320708.1 | 8 - 9 | 854 | 68 | ||
| NM_104980.4 | NP_176490.2 | 7 - 8 | 1019 | 68 | ||
| GenBank mRNA | AY826515.1 | - | 6 - 7 | 745 | 68 | |
| DQ020657.1 | - | 7 - 8 | 1019 | 68 | ||
More Information
Set Membership: |
|

NRPD1A
Genomic Map