Assay Details
| Gene Symbol: LPLAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104983.5 | NP_176493.1 | 5 - 6 | 1253 | 109 | |
| GenBank mRNA | AY072080.1 | - | 5 - 6 | 1160 | 109 | |
| AY122979.1 | - | 5 - 6 | 1076 | 109 | ||
More Information
Set Membership: |
|

LPLAT2
Genomic Map