Assay Details
| Gene Symbol: AT4G12060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117276.3 | NP_567386.1 | 1 - 2 | 334 | 79 | |
| GenBank mRNA | AK119030.1 | - | 1 - 2 | 282 | 79 | |
More Information
Set Membership: |
|

AT4G12060
Genomic Map