Assay Details
| Gene Symbol: AT4G12360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117306.3 | NP_192973.1 | 2 - 3 | 482 | 65 | |
| GenBank mRNA | BT010827.1 | - | 2 - 3 | 482 | 65 | |
| BT011305.1 | - | 2 - 3 | 394 | 65 | ||
More Information
Set Membership: |
|

AT4G12360
Genomic Map