Assay Details
| Gene Symbol: PME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117309.3 | NP_192976.1 | 1 | 918 | 150 | |
| GenBank mRNA | AY054507.1 | - | 1 | 751 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PME1| Gene Symbol: PME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117309.3 | NP_192976.1 | 1 | 918 | 150 | |
| GenBank mRNA | AY054507.1 | - | 1 | 751 | 150 | |
Set Membership: |
|