Assay Details
| Gene Symbol: EARLI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117318.3 | NP_192985.1 | 1 | 818 | 127 | |
| GenBank mRNA | AF412111.1 | - | 1 | 761 | 127 | |
| AK230253.1 | - | 1455 | 127 | |||
More Information
Set Membership: |
|

EARLI1
Genomic Map