Assay Details
| Gene Symbol: AIR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336523.1 | NP_001324697.1 | 32 - 33 | 4987 | 104 | |
| NM_129022.8 | NP_181015.7 | 32 - 33 | 4798 | 104 | ||
More Information
Set Membership: |
|

AIR9
Genomic Map