Assay Details
| Gene Symbol: FAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129030.2 | NP_181023.1 | 6 - 7 | 832 | 77 | |
| GenBank mRNA | AF021804.1 | - | 5 - 6 | 593 | 77 | |
| AY050326.1 | - | 6 - 7 | 633 | 77 | ||
| BT000482.1 | - | 5 - 6 | 593 | 77 | ||
More Information
Set Membership: |
|

FAH1
Genomic Map