Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 14 - 15 | 1781 | 86 | |
| NM_129064.3 | NP_181056.2 | 13 - 14 | 1747 | 86 | ||
| GenBank mRNA | AY496700.1 | - | 14 - 15 | 1740 | 86 | |
More Information
Set Membership: |
|

GRL
Genomic Map