Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 18 - 19 | 2311 | 64 | |
| NM_129064.3 | NP_181056.2 | 17 - 18 | 2277 | 64 | ||
| GenBank mRNA | AY496700.1 | - | 18 - 19 | 2270 | 64 | |
More Information
Set Membership: |
|

GRL
Genomic Map