Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 22 - 23 | 3892 | 78 | |
| NM_129064.3 | NP_181056.2 | 21 - 22 | 3858 | 78 | ||
| GenBank mRNA | AY496700.1 | - | 22 - 23 | 3851 | 78 | |
More Information
Set Membership: |
|

GRL
Genomic Map