Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 3 - 4 | 707 | 73 | |
| NM_129064.3 | NP_181056.2 | 3 - 4 | 586 | 73 | ||
| GenBank mRNA | AY496700.1 | - | 3 - 4 | 666 | 73 | |
More Information
Set Membership: |
|

GRL
Genomic Map