Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 8 - 9 | 1164 | 119 | |
| NM_129064.3 | NP_181056.2 | 8 - 9 | 1043 | 119 | ||
| GenBank mRNA | AY496700.1 | - | 8 - 9 | 1123 | 119 | |
More Information
Set Membership: |
|

GRL
Genomic Map