Assay Details
| Gene Symbol: GRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036408.3 | NP_001031485.1 | 12 - 13 | 1595 | 66 | |
| NM_129064.3 | NP_181056.2 | 11 - 12 | 1561 | 66 | ||
| GenBank mRNA | AY496700.1 | - | 12 - 13 | 1554 | 66 | |
More Information
Set Membership: |
|

GRL
Genomic Map