Assay Details
| Gene Symbol: EYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129084.3 | NP_565803.1 | 2 - 3 | 370 | 148 | |
| GenBank mRNA | AF410332.1 | - | 2 - 3 | 299 | 148 | |
| AY054555.1 | - | 2 - 3 | 324 | 148 | ||
| AY064618.1 | - | 1 - 2 | 238 | 148 | ||
| AY149930.1 | - | 1 - 2 | 238 | 148 | ||
More Information
Set Membership: |
|

EYA
Genomic Map