Assay Details
| Gene Symbol: SPPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105046.3 | NP_564815.1 | 2 - 3 | 787 | 66 | |
| NM_202353.1 | NP_974082.1 | 2 - 3 | 421 | 66 | ||
| GenBank mRNA | AY062807.1 | - | 2 - 3 | 374 | 66 | |
| BT008895.1 | - | 2 - 3 | 309 | 66 | ||
More Information
Set Membership: |
|

SPPL2
Genomic Map