Assay Details
| Gene Symbol: SPO11-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334143.1 | NP_001320320.1 | 1 - 2 | 116 | 75 | |
| NM_105072.3 | NP_176582.2 | 1 - 2 | 116 | 75 | ||
| GenBank mRNA | AJ251990.1 | - | 1 - 2 | 105 | 75 | |
| DQ446392.1 | - | 1 - 2 | 82 | 75 | ||
More Information
Set Membership: |
|

SPO11-2
Genomic Map