Assay Details
| Gene Symbol: SPO11-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334143.1 | NP_001320320.1 | 3 - 4 | 336 | 80 | |
| NM_105072.3 | NP_176582.2 | 3 - 4 | 336 | 80 | ||
| GenBank mRNA | AJ251990.1 | - | 3 - 4 | 325 | 80 | |
| DQ446392.1 | - | 3 - 4 | 302 | 80 | ||
More Information
Set Membership: |
|

SPO11-2
Genomic Map