Assay Details
| Gene Symbol: SPO11-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105072.3 | NP_176582.2 | 9 - 10 | 926 | 63 | |
| GenBank mRNA | AJ251990.1 | - | 9 - 10 | 915 | 63 | |
| DQ446392.1 | - | 9 - 10 | 892 | 63 | ||
More Information
Set Membership: |
|

SPO11-2
Genomic Map