Assay Details
| Gene Symbol: DAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124072.1 | NP_001117544.1 | 9 - 10 | 1301 | 80 | |
| NM_001334146.1 | NP_001319315.1 | 10 - 11 | 1532 | 80 | ||
| NM_001334147.1 | NP_001323399.1 | 9 - 10 | 1307 | 80 | ||
| NM_105085.2 | NP_564824.1 | 10 - 11 | 1489 | 80 | ||
| NM_179511.2 | NP_849842.1 | 10 - 11 | 1581 | 80 | ||
| GenBank mRNA | AY056097.1 | - | 10 - 11 | 1489 | 80 | |
| BT006360.1 | - | 9 - 10 | 1264 | 80 | ||
More Information
Set Membership: |
|

DAA1
Genomic Map