Assay Details
| Gene Symbol: DAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124072.1 | NP_001117544.1 | 6 - 7 | 645 | 69 | |
| NM_001334146.1 | NP_001319315.1 | 7 - 8 | 876 | 69 | ||
| NM_001334147.1 | NP_001323399.1 | 6 - 7 | 645 | 69 | ||
| NM_105085.2 | NP_564824.1 | 7 - 8 | 827 | 69 | ||
| NM_179511.2 | NP_849842.1 | 7 - 8 | 919 | 69 | ||
| GenBank mRNA | AY056097.1 | - | 7 - 8 | 827 | 69 | |
| BT006360.1 | - | 6 - 7 | 602 | 69 | ||
More Information
Set Membership: |
|

DAA1
Genomic Map