Assay Details
| Gene Symbol: RPN12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105127.4 | NP_176633.1 | 5 - 6 | 857 | 116 | |
| GenBank mRNA | AF410265.1 | - | 5 - 6 | 713 | 116 | |
| AY039857.1 | - | 5 - 6 | 676 | 116 | ||
| AY143888.1 | - | 5 - 6 | 667 | 116 | ||
| AY230846.1 | - | 5 - 6 | 690 | 116 | ||
More Information
Set Membership: |
|

RPN12a
Genomic Map