Assay Details
| Gene Symbol: AT1G64530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334166.1 | NP_001323413.1 | 3 - 4 | 853 | 125 | |
| NM_105128.5 | NP_176634.1 | 2 - 3 | 968 | 125 | ||
| GenBank mRNA | BT000933.1 | - | 2 - 3 | 775 | 125 | |
More Information
Set Membership: |
|

AT1G64530
Genomic Map