Assay Details
| Gene Symbol: AT1G64530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334166.1 | NP_001323413.1 | 5 - 6 | 1814 | 104 | |
| NM_105128.5 | NP_176634.1 | 4 - 5 | 1929 | 104 | ||
| GenBank mRNA | BT000933.1 | - | 4 - 5 | 1736 | 104 | |
More Information
Set Membership: |
|

AT1G64530
Genomic Map