Assay Details
| Gene Symbol: AT1G65220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105195.3 | NP_564845.1 | 6 - 7 | 1218 | 85 | |
| GenBank mRNA | AY056181.1 | - | 6 - 7 | 1035 | 85 | |
| BT006174.1 | - | 6 - 7 | 1035 | 85 | ||
More Information
Set Membership: |
|

AT1G65220
Genomic Map