Assay Details
| Gene Symbol: CBSX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105206.3 | NP_176711.1 | 1 - 2 | 704 | 92 | |
| GenBank mRNA | AK117112.1 | - | 1 - 2 | 657 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBSX6| Gene Symbol: CBSX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105206.3 | NP_176711.1 | 1 - 2 | 704 | 92 | |
| GenBank mRNA | AK117112.1 | - | 1 - 2 | 657 | 92 | |
Set Membership: |
|