Assay Details
| Gene Symbol: AT1G65370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105211.4 | NP_564849.1 | 2 - 3 | 569 | 121 | |
| GenBank mRNA | AY054271.1 | - | 2 - 3 | 365 | 121 | |
| AY132005.1 | - | 1 - 2 | 99 | 121 | ||
More Information
Set Membership: |
|

AT1G65370
Genomic Map