Assay Details
| Gene Symbol: AT1G65370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105211.4 | NP_564849.1 | 3 - 4 | 846 | 109 | |
| GenBank mRNA | AY054271.1 | - | 3 - 4 | 642 | 109 | |
| AY132005.1 | - | 2 - 3 | 376 | 109 | ||
More Information
Set Membership: |
|

AT1G65370
Genomic Map