Assay Details
| Gene Symbol: AT3G01015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_110965.4 | NP_186749.2 | 1 - 2 | 954 | 66 | |
| GenBank mRNA | AY735598.1 | - | 1 - 2 | 954 | 66 | |
| DQ056582.1 | - | 1 - 2 | 812 | 66 | ||
More Information
Set Membership: |
|

AT3G01015
Genomic Map