Assay Details
| Gene Symbol: AT3G01015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_110965.4 | NP_186749.2 | 4 - 5 | 1214 | 96 | |
| GenBank mRNA | AY735598.1 | - | 4 - 5 | 1214 | 96 | |
| DQ056582.1 | - | 4 - 5 | 1072 | 96 | ||
More Information
Set Membership: |
|

AT3G01015
Genomic Map