Assay Details
| Gene Symbol: HB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_110988.3 | NP_186771.1 | 2 - 3 | 813 | 124 | |
| GenBank mRNA | AK228890.1 | - | 2 - 3 | 634 | 124 | |
| BT029221.1 | - | 2 - 3 | 521 | 124 | ||
More Information
Set Membership: |
|

HB20
Genomic Map