Assay Details
| Gene Symbol: DEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_110999.7 | NP_186782.2 | 4 - 5 | 361 | 94 | |
| GenBank mRNA | AB074532.1 | - | 4 - 5 | 290 | 94 | |
| AY093024.1 | - | 4 - 5 | 320 | 94 | ||
| BT000412.1 | - | 4 - 5 | 290 | 94 | ||
More Information
Set Membership: |
|

DEL3
Genomic Map