Assay Details
| Gene Symbol: AIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117325.2 | NP_192992.1 | 1 | 557 | 104 | |
| GenBank mRNA | AF410287.1 | - | 1 | 557 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AIR1| Gene Symbol: AIR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117325.2 | NP_192992.1 | 1 | 557 | 104 | |
| GenBank mRNA | AF410287.1 | - | 1 | 557 | 104 | |
Set Membership: |
|