Assay Details
| Gene Symbol: CPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084908.2 | NP_001078377.1 | 1 - 2 | 1338 | 161 | |
| NM_001340790.1 | NP_001328191.1 | 1 - 2 | 1338 | 161 | ||
| NM_117326.4 | NP_192993.2 | 1 - 2 | 1338 | 161 | ||
More Information
Set Membership: |
|

CPR1
Genomic Map