Assay Details
| Gene Symbol: AT4G12710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117340.5 | NP_193007.1 | 1 - 2 | 461 | 71 | |
| GenBank mRNA | AK228965.1 | - | 1 - 2 | 433 | 71 | |
| BT026433.1 | - | 1 - 2 | 365 | 71 | ||
More Information
Set Membership: |
|

AT4G12710
Genomic Map